Learn More
5-Chloromethyl-4-methyl-2-(4-trifluoromethylphenyl)thiazole, TRC
Specifications
Specifications
| Chemical Name or Material | 5-Chloromethyl-4-methyl-2-(4-trifluoromethylphenyl)thiazole |
| CAS | 317318-97-1 |
| Molecular Formula | C12H9ClF3NS |
| Quantity | 20 mg |
| Synonym | 5-(Chloromethyl)-4-methyl-2-(4-trifluoromethylphenyl)-1,3-thiazole |
| InChI Formula | InChI=1S/C12H9ClF3NS/c1-7-10(6-13)18-11(17-7)8-2-4-9(5-3-8)12(14,15)16/h2-5H,6H2,1H3 |
| SMILES | Cc1nc(sc1CCl)c2ccc(cc2)C(F)(F)F |
| IUPAC Name | 5-(chloromethyl)-4-methyl-2-[4-(trifluoromethyl)phenyl]-1,3-thiazole |
| Molecular Weight (g/mol) | 291.72 |
| Formula Weight | 291.0096 |