Learn More
5-Chlorobenzo[d]thiazole, TRC
Product Code. 30367440
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
1g
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30367440 5 g 5g
30358637 1 g 1g
30363378 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30367440 Supplier Toronto Research Chemicals Supplier No. TRCC4290985G
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2786-51-8
Molecular Formula C7H4NSCl
Molecular Weight (g/mol) 169.63
Synonym 5-Chlorobenzothiazole
IUPAC Name 5-chloro-1,3-benzothiazole
SMILES Clc1ccc2scnc2c1
Specifications
Quantity 5 g
InChI Formula InChI=1S/C7H4ClNS/c8-5-1-2-7-6(3-5)9-4-10-7/h1-4H
Formula Weight 168.9753
Chemical Name or Material 5-Chlorobenzo[d]thiazole