Learn More
5-Chloro Hydrochlorothiazide, TRC
Product Code. 30010064
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30010064 25 mg 25mg
17984035 50 mg 50mg
17984105 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30010064 Supplier Toronto Research Chemicals Supplier No. TRCC36655025MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 5233-42-1
Molecular Formula C7 H7 Cl2 N3 O4 S2
Molecular Weight (g/mol) 332.18
Synonym 5-Chlorohydrochlorothiazide, 2H-1,2,4-Benzothiadiazine-7-sulfonamide, 5,6-dichloro-3,4-dihydro-, 1,1-dioxide, 5,6-Dichloro-3,4-dihydro-2H-1,2,4-benzothiadiazine-7-sulfonamide 1,1-dioxide
IUPAC Name 5,6-dichloro-1,1-dioxo-3,4-dihydro-2H-1λ^{6},2,4-benzothiadiazine-7-sulfonamide
SMILES NS(=O)(=O)c1cc2c(NCNS2(=O)=O)c(Cl)c1Cl
Specifications
Quantity 25 mg
InChI Formula InChI=1S/C7H7Cl2N3O4S2/c8-5-3(17(10,13)14)1-4-7(6(5)9)11-2-12-18(4,15)16/h1,11-12H,2H2,(H2,10,13,14)
Formula Weight 330.9255
Percent Purity >95%
Chemical Name or Material 5-Chloro Hydrochlorothiazide
Grade HPLC