Learn More
5-Chloro-2-oxindole, TRC
Product Code. 30358663
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30358663 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30358663 Supplier Toronto Research Chemicals Supplier No. TRCC3750301G
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 5-Chloro-2-indolinone
CAS 17630-75-0
Molecular Formula C8 H6 Cl N O
Quantity 1 g
Synonym 2H-Indol-2-one, 5-chloro-1,3-dihydro-, 2-Indolinone, 5-chloro- (8CI), Oxindole, 5-chloro- (6CI), 5-Chloro-1,3-dihydro-2H-indol-2-one, 5-Chloro-1,3-dihydroindol-2-one, 5-Chloro-2-oxindole, 5-Chloro-2-oxoindoline, 5-Chloroindol-2-one, 5-Chlorooxindole
InChI Formula InChI=1S/C8H6ClNO/c9-6-1-2-7-5(3-6)4-8(11)10-7/h1-3H,4H2,(H,10,11)
SMILES Clc1ccc2NC(=O)Cc2c1
IUPAC Name 5-chloro-1,3-dihydroindol-2-one
Molecular Weight (g/mol) 167.592
Formula Weight 167.0138