Learn More
5-Chloro-2-oxindole, TRC
Specifications
Specifications
| Chemical Name or Material | 5-Chloro-2-indolinone |
| CAS | 17630-75-0 |
| Molecular Formula | C8 H6 Cl N O |
| Quantity | 2.5 g |
| Synonym | 2H-Indol-2-one, 5-chloro-1,3-dihydro-, 2-Indolinone, 5-chloro- (8CI), Oxindole, 5-chloro- (6CI), 5-Chloro-1,3-dihydro-2H-indol-2-one, 5-Chloro-1,3-dihydroindol-2-one, 5-Chloro-2-oxindole, 5-Chloro-2-oxoindoline, 5-Chloroindol-2-one, 5-Chlorooxindole |
| InChI Formula | InChI=1S/C8H6ClNO/c9-6-1-2-7-5(3-6)4-8(11)10-7/h1-3H,4H2,(H,10,11) |
| SMILES | Clc1ccc2NC(=O)Cc2c1 |
| IUPAC Name | 5-chloro-1,3-dihydroindol-2-one |
| Molecular Weight (g/mol) | 167.592 |
| Formula Weight | 167.0138 |