missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-fluorobenzeneboronic acid, 97%

Product Code. p-7035391
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15429877

Brand: Thermo Scientific Alfa Aesar H53108.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 352535-83-2 | C6H5BClFO2 | 174.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 352535-83-2
Molecular Formula C6H5BClFO2
Molecular Weight (g/mol) 174.36
MDL Number MFCD05664225
InChI Key GGTUVWGMCFXUAS-UHFFFAOYSA-N
Synonym 5-chloro-2-fluorophenyl boronic acid, 5-chloro-2-fluorobenzeneboronic acid, 5-chloro-2-fluorophenylboronicacid, 5-chloro-2-fluoro-phenyl boronic acid, boronic acid, 5-chloro-2-fluorophenyl, pubchem11819, acmc-1ae0a, boronic acid,b-5-chloro-2-fluorophenyl, boronic acid, b-5-chloro-2-fluorophenyl
PubChem CID 3718925
IUPAC Name (5-chloro-2-fluorophenyl)boronic acid
SMILES OB(O)C1=CC(Cl)=CC=C1F

Specifications

Melting Point 182°C to 188°C
Quantity 1 g
Formula Weight 174.37
Percent Purity 97%
Chemical Name or Material 5-Chloro-2-fluorobenzeneboronic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.