Learn More
5-Chloro-2-(3,4-dimethoxyphenyl)-2-isopropylvaleronitrile, TRC
Specifications
Specifications
| Chemical Name or Material | 5-Chloro-2-(3,4-dimethoxyphenyl)-2-isopropylvaleronitrile |
| CAS | 27487-83-8 |
| Molecular Formula | C16 H22 Cl N O2 |
| Quantity | 100 mg |
| InChI Formula | InChI=1S/C16H22ClNO2/c1-12(2)16(11-18,8-5-9-17)13-6-7-14(19-3)15(10-13)20-4/h6-7,10,12H,5,8-9H2,1-4H3 |
| SMILES | COc1ccc(cc1OC)C(CCCCl)(C#N)C(C)C |
| IUPAC Name | 5-chloro-2-(3,4-dimethoxyphenyl)-2-propan-2-ylpentanenitrile |
| Molecular Weight (g/mol) | 295.8 |
| Formula Weight | 295.1339 |