missing translation for 'onlineSavingsMsg'
Learn More

5-Bromotryptamine hydrochloride, 98%, Thermo Scientific™

Product Code. 11318726
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11318726 250 mg 250mg
11308726 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11318726 Supplier Thermo Scientific Alfa Aesar Supplier No. L19418.MD
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
5-Bromotryptamine hydrochloride is used as pharmaceutical intermediates.

Notes
Light sensitive. Store in cool dry conditions. Incompatible with light and oxidizing agents. Store in the dark.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 81868-12-4
Molecular Formula C10H12BrClN2
Molecular Weight (g/mol) 275.574
MDL Number MFCD00152022
InChI Key QUPXCOYSKSUJPX-UHFFFAOYSA-N
Synonym 2-5-bromo-1h-indol-3-yl ethanamine hydrochloride, 5-bromotryptamine hydrochloride, 2-5-bromo-1h-indol-3-yl-ethylamine hydrochloride, 1h-indole-3-ethanamine, 5-bromo-, monohydrochloride, 5-bromotryptamine hcl, 2-5-bromo-1h-indol-3-yl ethanaminehydrochloride, 2-5-bromo-1h-indol-3-yl ethan-1-amine hydrochloride, 2-5-bromo-1h-indol-3-yl ethan-1-amine-hydrogen chloride 1/1
PubChem CID 13241175
IUPAC Name 2-(5-bromo-1H-indol-3-yl)ethanamine;hydrochloride
SMILES C1=CC2=C(C=C1Br)C(=CN2)CCN.Cl

Specifications

Melting Point ∽274°C (decomposition)
Quantity 250 mg
Beilstein 3728224
Sensitivity Light sensitive
Formula Weight 275.58
Percent Purity 98%
Chemical Name or Material 5-Bromotryptamine hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.