Learn More
(5-Bromopyridin-2-yl)[2-(tert-butyldimethylsilyloxy)ethyl]methylamine, TRC
Specifications
Specifications
| Chemical Name or Material | (5-Bromopyridin-2-yl)[2-(tert-butyldimethylsilyloxy)ethyl]me |
| CAS | 326496-01-9 |
| Molecular Formula | C14 H25 Br N2 O Si |
| Quantity | 500 mg |
| InChI Formula | InChI=1S/C14H25BrN2OSi/c1-14(2,3)19(5,6)18-10-9-17(4)13-8-7-12(15)11-16-13/h7-8,11H,9-10H2,1-6H3 |
| SMILES | CN(CCO[Si](C)(C)C(C)(C)C)c1ccc(Br)cn1 |
| IUPAC Name | 5-bromo-N-[2-[tert-butyl(dimethyl)silyl]oxyethyl]-N-methylpyridin-2-amine |
| Molecular Weight (g/mol) | 345.35 |
| Formula Weight | 344.092 |