Learn More
5-Bromo-6-chlorobenzene-1,2,3,4-d4, TRC
Product Code. 30329881
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30329881
Brand: Toronto Research Chemicals TRCB682312100MG
Please to purchase this item. Need a web account? Register with us today!
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 5-Bromo-6-chlorobenzene-1,2,3,4-d4
CAS 1219795-51-3
Molecular Formula C6 D4 Br Cl
Quantity 100 mg
Synonym 2-Bromochlorobenzene-d4, Benzene-1,2,3,4-d4, 5-bromo-6-chloro- (ACI), 5-Bromo-6-chlorobenzene-1,2,3,4-d4 (ACI)
InChI Formula InChI=1S/C6H4BrCl/c7-5-3-1-2-4-6(5)8/h1-4H/i1D,2D,3D,4D
SMILES [2H]c1c([2H])c([2H])c(Br)c(Cl)c1[2H]
IUPAC Name 1-bromo-2-chloro-3,4,5,6-tetradeuteriobenzene
Molecular Weight (g/mol) 195.48
Formula Weight 193.9436