Learn More
5-Bromo-4-(bromomethyl)-1-pentene (Stabilized with Copper wire), TRC
Product Code. 30338219
Change view
Click to view available options
Quantity:
250 mg
500 mg
Unit Size:
250mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30338219 500 mg 500mg
30331684 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30338219 Supplier Toronto Research Chemicals Supplier No. TRCB577490500MG
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 5-Bromo-4-bromomethyl-1-pentene
CAS 157556-86-0
Molecular Formula C6H10Br2
Quantity 500 mg
Synonym 5-bromo-4-(bromomethyl)pent-1-ene
InChI Formula InChI=1S/C6H10Br2/c1-2-3-6(4-7)5-8/h2,6H,1,3-5H2
SMILES BrCC(CBr)CC=C
IUPAC Name 5-bromo-4-(bromomethyl)pent-1-ene
Molecular Weight (g/mol) 241.95
Formula Weight 239.91