Learn More
5-Bromo-2-(N-methylamino)pyridine, TRC
Specifications
Specifications
| Chemical Name or Material | 5-Bromo-2-(N-methylamino)pyridine |
| CAS | 84539-30-0 |
| Molecular Formula | C6H7BrN2 |
| Quantity | 100 mg |
| InChI Formula | InChI=1S/C6H7BrN2/c1-8-6-3-2-5(7)4-9-6/h2-4H,1H3,(H,8,9) |
| SMILES | CNc1ccc(Br)cn1 |
| IUPAC Name | 5-bromo-N-methylpyridin-2-amine |
| Molecular Weight (g/mol) | 187.04 |
| Formula Weight | 185.9793 |