missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-fluoroaniline, 98%, Thermo Scientific Chemicals

Catalog Number p-7053913
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
2924-09-6
C6H5BrFN
190.02
MFCD03094181
ADWKOCXRCRSMLQ-UHFFFAOYSA-N
This item is not returnable. View return policy
2924-09-6
C6H5BrFN
190.02
MFCD03094181
ADWKOCXRCRSMLQ-UHFFFAOYSA-N

5-Bromo-2-fluoroaniline is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Bromo-2-fluoroaniline is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 27°C
Boiling Point 105°C (12 mmHg)
Quantity 25 g
UN Number UN2811
Beilstein 1241528
Synonym 2,4-diisopropylphenol, phenol, 2,4-bis 1-methylethyl, phenol, 2,4-diisopropyl, 2,4-di propan-2-yl phenol, 2,4-diisopropyl-phenol, unii-yd41xsg2l8, yd41xsg2l8, 2,4-bis 1-methylethyl phenol, propofol impurity a, acmc-20aocr
Solubility Information Slightly soluble in water.
SMILES NC1=C(F)C=CC(Br)=C1
IUPAC Name 2,4-di(propan-2-yl)phenol
PubChem CID 18048
Formula Weight 190.01
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 5-Bromo-2-fluoroaniline
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'