missing translation for 'onlineSavingsMsg'
Learn More

5-Aminomethyl-2-(Boc-amino)pyridine, 97%

Product Code. p-7033068
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11454490 1 g 1g
11464490 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11454490 Supplier Thermo Scientific Alfa Aesar Supplier No. H50078.03
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 187237-37-2 | C11H17N3O2 | 223.276 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 187237-37-2
Molecular Formula C11H17N3O2
Molecular Weight (g/mol) 223.276
MDL Number MFCD07781132
InChI Key DFLQTVPEIMTXSZ-UHFFFAOYSA-N
Synonym 2-boc-amino-5-aminomethyl pyridine, 5-aminomethyl-2-n-boc aminopyridine, tert-butyl 5-aminomethyl pyridin-2-yl carbamate, tert-butyl 5-aminomethyl pyridin-2-ylcarbamate, tert-butyl n-5-aminomethyl pyridin-2-yl carbamate, 5-aminomethyl-pyridin-2-yl-carbamic acid tert-butyl ester, 5-aminomethyl-2-boc-amino pyridine, carbamic acid,n-5-aminomethyl-2-pyridinyl-, 1,1-dimethylethyl ester, 2-tert-butoxycarbonyl-amino-5-aminomethylpyridine
PubChem CID 42553118
IUPAC Name tert-butyl N-[5-(aminomethyl)pyridin-2-yl]carbamate
SMILES CC(C)(C)OC(=O)NC1=NC=C(C=C1)CN

Specifications

Melting Point 132°C to 134°C
Quantity 1 g
Sensitivity Air Sensitive
Formula Weight 223.28
Percent Purity 97%
Chemical Name or Material 5-Aminomethyl-2-(Boc-amino)pyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.