missing translation for 'onlineSavingsMsg'
Learn More

5-Aminoimidazole-4-carboxamide 1-beta-D-ribofuranoside, 98%, Thermo Scientific Chemicals

Product Code. 15440387
Change view
Click to view available options
Quantity:
50 mg
200 mg
Unit Size:
200mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15440387 200 mg 200mg
15450387 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15440387 Supplier Thermo Scientific Alfa Aesar Supplier No. J64720.!3
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Activator of AMP-activated protein kinase (AMPK)

5-Aminoimidazole-4-carboxamide 1-beta-D-ribofuranoside is used as a cell permeable activator of AMP-activated protein kinase (AMPK), a metabolic master regulator that is activated in times of reduced energy availability (high cellular AMP:ATP ratios) and serves to inhibit anabolic processes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Aminoimidazole-4-carboxamide 1-beta-D-ribofuranoside is used as a cell permeable activator of AMP-activated protein kinase (AMPK), a metabolic master regulator that is activated in times of reduced energy availability (high cellular AMP:ATP ratios) and serves to inhibit anabolic processes. In vivo, pharmacologic activation of AMPK with AICAR mimics exercise and triggers insulin-independent glucose uptake by skeletal muscle.

Solubility
Soluble in DMSO at 2mg/ml. Soluble in water or ethanol at less than 1mg/ml

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2627-69-2
Molecular Formula C9H14N4O5
Molecular Weight (g/mol) 258.23
MDL Number MFCD00869751
InChI Key RTRQQBHATOEIAF-UUOKFMHZSA-N
Synonym acadesine, aica-riboside, aicar, arasine, aica riboside, aic-riboside, acadesina, acadesinum, 5-amino-1-beta-d-ribofuranosyl-1h-imidazole-4-carboxamide, protara
PubChem CID 17513
ChEBI CHEBI:28498
IUPAC Name 5-amino-1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]imidazole-4-carboxamide
SMILES NC(=O)C1=C(N)N(C=N1)[C@@H]1O[C@H](CO)[C@@H](O)[C@H]1O

Specifications

Melting Point 214°C to 215°C
Color Brown
Quantity 200 mg
Merck Index 14,16
Solubility Information Soluble in DMSO at 2mg/ml; Soluble in water or ethanol at less than 1mg/ml
Formula Weight 258.23
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 5-Aminoimidazole-4-carboxamide 1-beta-D-ribofuranoside

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.