missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-2-bromo-3-fluoropyridine, 97%, Thermo Scientific™

Product Code. 15435957
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15435957 250 mg 250mg
15415957 1 g 1g
15425957 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15435957 Supplier Thermo Scientific Alfa Aesar Supplier No. H33154.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Commercially available 5-bromo-2-fluoropyridine was treated with LDA at -78°C to give the lithiated species. Fluoropyridine-based bromoacetamide reagent has been prepared and used in prosthetic group introduction for the labeling of oligonucleotides.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents. Refrigerate(2-7°C ).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1256276-41-1
Molecular Formula C5H4BrFN2
Molecular Weight (g/mol) 191.003
MDL Number MFCD21099397
InChI Key DDJLNVAJTGIWNJ-UHFFFAOYSA-N
Synonym 5-amino-2-bromo-3-fluoropyridine, 3-pyridinamine, 6-bromo-5-fluoro
PubChem CID 71741382
IUPAC Name 6-bromo-5-fluoropyridin-3-amine
SMILES C1=C(C=NC(=C1F)Br)N

Specifications

Melting Point 114°C to 118°C
Quantity 250 mg
Formula Weight 191
Percent Purity 97%
Chemical Name or Material 5-Amino-2-bromo-3-fluoropyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.