Learn More
5,8-Dihydro-1-naphthol (>80%), TRC
Specifications
Specifications
| Chemical Name or Material | 5,8-Dihydro-1-naphthol |
| CAS | 27673-48-9 |
| Molecular Formula | C10 H10 O |
| Quantity | 1 g |
| Synonym | 1-Naphthalenol, 5,8-dihydro-, 1-Naphthol, 5,8-dihydro- (6CI,8CI), 5,8-Dihydro-1-naphthalenol, 5,8-Dihydro-1-naphthol, 5,8-Dihydronaphthalen-1-ol, 5-Hydroxy-1,4-dihydronaphthalene, NSC 125584 |
| InChI Formula | InChI=1S/C10H10O/c11-10-7-3-5-8-4-1-2-6-9(8)10/h1-3,5,7,11H,4,6H2 |
| SMILES | Oc1cccc2CC=CCc12 |
| IUPAC Name | 5,8-dihydronaphthalen-1-ol |
| Molecular Weight (g/mol) | 146.19 |
| Formula Weight | 146.0732 |