Learn More
5,6-Dihydroxyindole, TRC
Specifications
Specifications
| Chemical Name or Material | 5,6-Dihydroxyindole |
| CAS | 3131-52-0 |
| Molecular Formula | C8H7NO2 |
| Quantity | 500 mg |
| Synonym | Dopamine Impurity 22 |
| InChI Formula | InChI=1S/C8H7NO2/c10-7-3-5-1-2-9-6(5)4-8(7)11/h1-4,9-11H |
| SMILES | Oc1cc2cc[nH]c2cc1O |
| Molecular Weight (g/mol) | 149.15 |
| Formula Weight | 149.0477 |