Learn More
5,6-Diamino-1,3-dipropyl-2,4(1H,3H)-Pyrimidinedione Monohydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | 5,6-Diamino-1,3-dipropyl-2,4(1H,3H)-Pyrimidinedione Monohydrochloride |
| CAS | 324002-49-5 |
| Molecular Formula | C10H18N4O2 . (HCl) |
| Quantity | 1 g |
| Synonym | 2,4(1H,3H)-Pyrimidinedione, 5,6-diamino-1,3-dipropyl-, hydrochloride (1:1) (ACI), 2,4(1H,3H)-Pyrimidinedione, 5,6-diamino-1,3-dipropyl-, monohydrochloride (9CI), 5,6-Diamino-1,3-dipropyl-2,4(1H,3H)-pyrimidinedione monohydrochloride, 1,3-Dipropyl-5,6-diaminouracil monohydrochloride, 5,6-Diamino-1,3-dipropyl-1,3-dihydropyrimidine-2,4-dione monohydrochloride, 5,6-Diamino-1,3-dipropyl-1H-pyrimidine-2,4-dione monohydrochloride, 5,6-Diamino-1,3-dipropyluracil monohydrochloride |
| InChI Formula | InChI=1S/C10H18N4O2.ClH/c1-3-5-13-8(12)7(11)9(15)14(6-4-2)10(13)16;/h3-6,11-12H2,1-2H3;1H |
| SMILES | Cl.CCCC1C(=C(N)C(=O)C(CCC)C1=O)N |
| Molecular Weight (g/mol) | 262.74 |
| Formula Weight | 262.1197 |