missing translation for 'onlineSavingsMsg'
Learn More

5,6,7,8-Tetrahydro-1-naphthol, 99%

Product Code. p-7043737
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11307705 10 g 10g
11317705 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11307705 Supplier Thermo Scientific Alfa Aesar Supplier No. L02416.09
Add to basket
This item is not returnable. View return policy

CAS: 529-35-1 | C10H12O | 148.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 529-35-1
Molecular Formula C10H12O
Molecular Weight (g/mol) 148.21
MDL Number MFCD00001734
InChI Key SCWNNOCLLOHZIG-UHFFFAOYSA-N
Synonym 5,6,7,8-tetrahydro-1-naphthol, 5-hydroxytetralin, 1-naphthalenol, 5,6,7,8-tetrahydro, tetralin-5-ol, 5,6,7,8-tetrahydro-alpha-naphthol, unii-1r7b5i98hv, 5,6,7,8-tetrahydronaphthol, 5,6,7,8-tetrahydro-1-naphthalenol, 5,6,7,8-tetrahydro-naphthalen-1-ol, tetrahydro-.alpha.-naphthol
PubChem CID 68258
ChEBI CHEBI:45900
IUPAC Name 5,6,7,8-tetrahydronaphthalen-1-ol
SMILES OC1=CC=CC2=C1CCCC2

Specifications

Melting Point 69°C to 71°C
Boiling Point 264°C to 265°C (705 mmHg)
Quantity 10 g
Beilstein 1865081
Formula Weight 148.21
Percent Purity 99%
Chemical Name or Material 5,6,7,8-Tetrahydro-1-naphthol

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.