Learn More
5,5-Dimethyldihydrofuran-2-one, TRC
Specifications
Specifications
| Chemical Name or Material | 5,5-Dimethyldihydrofuran-2(3H)-one |
| CAS | 3123-97-5 |
| Molecular Formula | C6 H10 O2 |
| Quantity | 250 mg |
| Synonym | 2(3H)-Furanone, dihydro-5,5-dimethyl-, Valeric acid, 4-hydroxy-4-methyl-, lactone (6CI), Valeric acid, 4-hydroxy-4-methyl-, γ-lactone (7CI), Dihydro-5,5-dimethyl-2(3H)-furanone, 4,4-Dimethyl-4-butanolide, 4,4-Dimethylbutyrolactone, 4-Methyl-4-hydroxypentan |
| InChI Formula | InChI=1S/C6H10O2/c1-6(2)4-3-5(7)8-6/h3-4H2,1-2H3 |
| SMILES | CC1(C)CCC(=O)O1 |
| IUPAC Name | 5,5-dimethyloxolan-2-one |
| Molecular Weight (g/mol) | 114.142 |
| Formula Weight | 114.068 |