Learn More
5-(4-Pyridyl)-1H-tetrazole, 98%, Thermo Scientific Chemicals
Product Code. 11445606
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. 11445606
Brand: Thermo Scientific Alfa Aesar B20741.03
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 14389-12-9
Molecular Formula C6H5N5
Molecular Weight (g/mol) 147.141
MDL Number MFCD00068115
InChI Key JMILUUVWLRKJFB-UHFFFAOYSA-N
Synonym 5-4-pyridyl-1h-tetrazole, 4-2h-tetrazol-5-yl pyridine, 4-1h-tetrazol-5-yl pyridine, 4-2h-tetrazol-5-yl-pyridine, 4-1h-1,2,3,4-tetrazol-5-yl pyridine, 4-2h-1,2,3,4-tetrazol-5-yl pyridine, pyridine, 4-2h-tetrazol-5-yl, 5-4-pyridyl-1 h-tetrazole, 5-4-pyridyl-2h-1,2,3,4-tetraazole, acmc-20amxi
PubChem CID 482249
IUPAC Name 4-(2H-tetrazol-5-yl)pyridine
SMILES C1=CN=CC=C1C2=NNN=N2
Specifications
Quantity 1 g
Formula Weight 147.14
Percent Purity 98%
Chemical Name or Material 5-(4-Pyridyl)-1H-tetrazole