Learn More
5-(4-aminophenyl)-1,3,4-oxadiazole-2-thiol, TRC
Specifications
Specifications
| Chemical Name or Material | 5-(4-aminophenyl)-1,3,4-oxadiazole-2-thiol |
| CAS | 32058-82-5 |
| Molecular Formula | C8H7N3OS |
| Quantity | 500 mg |
| Synonym | 1,3,4-Oxadiazole-2(3H)-thione, 5-(4-aminophenyl)- (9CI, ACI), 1,3,4-Oxadiazole-2-thiol, 5-(p-aminophenyl)- (8CI), 5-(4-Aminophenyl)-1,3,4-oxadiazole-2(3H)-thione (ACI), δ2-1,3,4-Oxadiazoline-5-thione, 2-(p-aminophenyl)- (7CI), 2-(4-Aminophenyl)-1,3,4-oxadiazoline-5-thione, 2-(p-Aminophenyl)-1,3,4-oxadiazoline-5-thione, 5-(4-Aminophenyl)-1,3,4-oxadiazole-2-thiol, 5-(p-Aminophenyl)-1,3,4-oxadiazole-2-(3H)-thione |
| InChI Formula | InChI=1S/C8H7N3OS/c9-6-3-1-5(2-4-6)7-10-11-8(13)12-7/h1-4H,9H2,(H,11,13) |
| SMILES | Nc1ccc(cc1)C2=NNC(=S)O2 |
| IUPAC Name | 5-(4-aminophenyl)-3H-1,3,4-oxadiazole-2-thione |
| Molecular Weight (g/mol) | 193.22 |
| Formula Weight | 193.031 |