Learn More
5-(3-hydroxyphenyl)pentanoic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | 5-(3-hydroxyphenyl)pentanoic Acid |
| CAS | 31129-95-0 |
| Molecular Formula | C11H14O3 |
| Quantity | 100 mg |
| InChI Formula | InChI=1S/C11H14O3/c12-10-6-3-5-9(8-10)4-1-2-7-11(13)14/h3,5-6,8,12H,1-2,4,7H2,(H,13,14) |
| SMILES | OC(=O)CCCCc1cccc(O)c1 |
| IUPAC Name | 5-(3-hydroxyphenyl)pentanoic acid |
| Molecular Weight (g/mol) | 194.23 |
| Formula Weight | 194.0943 |