missing translation for 'onlineSavingsMsg'
Learn More

5-(3-Bromophenyl)-1-methyl-1H-pyrazole, 97%, Thermo Scientific™

Product Code. 10555564
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10555564 5 g Amber Glass Bottle 5g
10162944 250 mg Amber Glass Bottle 250mg
10708265 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10555564 Supplier Thermo Scientific Maybridge Supplier No. CC60510DE

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 425379-69-7
Molecular Formula C10H9BrN2
Molecular Weight (g/mol) 237.1
MDL Number MFCD09065005
InChI Key RIYUQLBSAMXENJ-UHFFFAOYSA-N
Synonym 5-3-bromophenyl-1-methyl-1h-pyrazole, 5-3-bromophenyl-1-methylpyrazole, 1h-pyrazole,5-3-bromophenyl-1-methyl
PubChem CID 22329560
IUPAC Name 5-(3-bromophenyl)-1-methylpyrazole
SMILES CN1C(=CC=N1)C2=CC(=CC=C2)Br

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 237.1
Percent Purity 97%
Chemical Name or Material 5-(3-Bromophenyl)-1-methyl-1H-pyrazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.