Learn More
5-(2-Hydroxyethyl)-4-methylthiazol-2(3H)-one, TRC
Specifications
Specifications
| Chemical Name or Material | 5-(2-Hydroxyethyl)-4-methylthiazol-2(3H)-one |
| CAS | 32795-64-5 |
| Molecular Formula | C6H9NO2S |
| Quantity | 10 mg |
| Synonym | 5-(2-hydroxyethyl)-4-methyl-2(3H)-Thiazolone |
| InChI Formula | InChI=1S/C6H9NO2S/c1-4-5(2-3-8)10-6(9)7-4/h8H,2-3H2,1H3,(H,7,9) |
| SMILES | O=C1SC(=C(N1)C)CCO |
| IUPAC Name | 5-(2-hydroxyethyl)-4-methylthiazol-2(3H)-one |
| Molecular Weight (g/mol) | 159.21 |
| Formula Weight | 159.04 |