Learn More
5-(1-Pyrrolidinyl)-2-furaldehyde, 98%, Thermo Scientific™
Product Code. 11419480
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. 11419480
Brand: Thermo Scientific Alfa Aesar H50316.MD
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 84966-28-9
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.19
MDL Number MFCD02629619
InChI Key TZLGFEAULCWLDT-UHFFFAOYSA-N
Synonym 5-pyrrolidin-1-yl furan-2-carbaldehyde, 5-pyrrolidin-1-yl-2-furaldehyde, 5-1-pyrrolidino-2-furaldehyde, 5-pyrrolidin-1-yl-furan-2-carbaldehyde, 5-1-pyrrolidinyl-2-furancarboxaldehyde, 5-1-pyrrolidinyl-2-furaldehyde, 5-pyrrolidinylfuran-2-carbaldehyde, acmc-20a6x5, 5-pyrrolidin-1-yl-2-furaldehyde saltdata: free
PubChem CID 801481
IUPAC Name 5-pyrrolidin-1-ylfuran-2-carbaldehyde
SMILES O=CC1=CC=C(O1)N1CCCC1
Specifications
Melting Point 82°C to 84°C
Quantity 250 mg
Formula Weight 165.19
Percent Purity 98%
Chemical Name or Material 5-(1-Pyrrolidinyl)-2-furaldehyde