Learn More
4H-Thieno[3,2-c]chromene-2-carboxylic acid, TRC
Specifications
Specifications
| Chemical Name or Material | 4H-Thieno[3,2-c]chromene-2-carboxylic acid |
| CAS | 26268-04-2 |
| Molecular Formula | C12H8O3S |
| Quantity | 50 mg |
| InChI Formula | InChI=1S/C12H8O3S/c13-12(14)10-5-7-6-15-9-4-2-1-3-8(9)11(7)16-10/h1-5H,6H2,(H,13,14) |
| SMILES | OC(=O)c1cc2COc3ccccc3c2s1 |
| IUPAC Name | 4H-thieno[3,2-c]chromene-2-carboxylic acid |
| Molecular Weight (g/mol) | 232.25 |
| Formula Weight | 232.0194 |