Learn More
4Alpha-Phorbol 12-Myristate 13-Acetate, TRC
Specifications
Specifications
| Chemical Name or Material | 4Alpha-Phorbol 12-Myristate 13-Acetate |
| CAS | 63597-44-4 |
| Molecular Formula | C36H56O8 |
| Quantity | 2.5 mg |
| Synonym | 1a,1b,4,4a,5,7a,7b,8,9,9a-Decahydro-4a,7b-dihydroxy-3-(1aR,1bS,4aS,7aS,7bS,8R,9R,9aS)-9a-(acetyloxy)-tetradecanoic Acid (Hydroxymethyl)-1,1,6,8-tetramethyl-5-oxo-1H-cyclopropa[3,4]benz[1,2-e]azulen-9-yl Ester, [1aR-(1aα,1bβ,4aα,7aα,7bα,8α,9β,9aα)]- 9a-(acetyloxy)-1a,1b,4,4a,5,7a,7b,8,9,9a-Decahydro-4a,7b-dihydroxy-3-(hydroxymethyl)-1,1,6,8-tetramethyl-5-oxo-1H-cyclopropa[3,4]benz[1,2-e]azulen-9-yl Ester Tetradecanoic Acid, 1H-Cyclopropa[3,4]benz[1,2-e]azulene','Tetradecanoic Acid Deriv., 4α-12-O-Tetradecanoylphorbol 13-Acetate, 4α-Phorbol 12-Myristate 13-Acetate, 4α-TPA, α-TPA |
| InChI Formula | InChI=1S/C36H56O8/c1-7-8-9-10-11-12-13-14-15-16-17-18-29(39)43-32-24(3)35(42)27(30-33(5,6)36(30,32)44-25(4)38)20-26(22-37)21-34(41)28(35)19-23(2)31(34)40/h19-20,24,27-28,30,32,37,41-42H,7-18,21-22H2,1-6H3/t24-,27+,28-,30-,32-,34+,35-,36-/m1/s1 |
| SMILES | CC1=C[C@@]2([H])[C@@]3(O)[C@H](C)[C@@H](OC(CCCCCCCCCCCCC)=O)[C@]4(OC(C)=O)[C@](C4(C)C)([H])[C@]3([H])C=C(CO)C[C@@]2(O)C1=O |
| IUPAC Name | (1aR,1bS,4aS,7aS,7bS,8R,9R,9aS)-9a-acetoxy-4a,7b-dihydroxy-3-(hydroxymethyl)-1,1,6,8-tetramethyl-5-oxo-1a,1b,4,4a,5,7a,7b,8,9,9a-decahydro-1H-cyclopropa[3,4]benzo[1,2-e]azulen-9-yl tetradecanoate |
| Molecular Weight (g/mol) | 616.83 |
| Formula Weight | 616.4 |