Learn More
KleenGuard™ G35 Seamless Knit Nylon Inspection Gloves
Product Code. 15776252
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
Unit Size:
Pack of 10
Pack of 240
Product Code. 15776252
Brand: KleenGuard™ 38718
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Low-lint gloves are ideal for detecting imperfections and defects when conducting critical quality control inspections. Ambidextrous KleenGuard™ G35 Seamless Knit Nylon Inspection Gloves are silicone-free for use in a wide variety of applications including film handling, aerospace, and automotive assembly.
  • Ambidextrous design allows gloves to be replaced one at a time, rather than in pairs
  • High degree of sensitivity and dexterity for inspecting for defects and imperfections
  • Designed for critical quality control inspections
  • Seamless knit nylon shell reduces skin irritation
  • Low lint for cleanroom applications
  • Silicone-free
Specifications
Construction Seamless Knit
Coating Material Uncoated
Color White
Disposable Single-use
Hand Ambidextrous
Latex Free Yes
Material Nylon
Product Type Inspection Gloves
Size Medium
Sterility Non-sterile
For Use With (Application) Automotive Assembly, Aerospace Engineering, and Quality Control Inspections
Glove Cuff Style Knit
Series KleenGuard™ G35