Learn More
42-(2-Tetrazolyl)rapamycin, MedChemExpress
Description
42-(2-Tetrazolyl)rapamycin is a prodrug compound of a rapamycin analog extracted from patent US 20080171763 A1, Example 1. Rapamycin is a specific mTOR inhibitor.
Specifications
Specifications
| Chemical Name or Material | 42-(2-Tetrazolyl)rapamycin |
| Physical Form | Solid |
| CAS | 221877-56-1 |
| Color | Off-White |
| Quantity | 5 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C52H79N5O12 |
| For Use With (Application) | Cancer-programmed cell death |
| Solubility Information | DMSO : ≥ 130 mg/mL (134.55 mM) |
| SMILES | [H][C@]1(C(O[C@@]([C@@H](C[C@H]2C[C@@H](OC)[C@@H](N3N=CN=N3)CC2)C)([H])CC([C@@H](/C=C([C@@H](O)[C@H]4OC)\C)C)=O)=O)N(C(C([C@@]5(O)[C@H](C)CC[C@](C[C@@H](/C(C)=C/C=C/C=C/[C@@H](C)C[C@@H](C)C4=O)OC)([H])O5)=O)=O)CCCC1 |
| Show More |