Learn More
B Braun™ Needle-free IV Injection Port Valve
Product Code. 10449569
Change view
Click to view available options
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. unitSize
10449569 Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10449569 Supplier B Braun™ Supplier No. 415111
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This item is not returnable. View return policy
B Braun™ Needle-free IV Injection Port Valve provides easy Luer Lock and Luer Slip access for aspiration, injection or gravity feed of IV fluids.
  • Lipid-resistant material
  • Flow rates up to 200mL/min.
  • 0.05mL positive displacement
  • Compatible with Luer Lock and Luer Slip, valve opens and closes automatically with each Luer taper
  • Not made with DEHP, PVC, or natural rubber latex
  • Simplifies handling
  • Helps prevent bloodstream infections
  • Helps reduce catheter occlusions
Specifications
Max. Pressure 300 psi