Learn More
RETSCH 400 dia. x 65mmH Stainless Steel Test Sieve, ISO Certified
Brand: RETSCH 60.166.000036
650.75 EUR valid until 2024-12-31
Use promo code "23265" to get your promotional price.
Additional Details : Weight : 1.60000kg
Specifications
Stainless Steel | |
Stainless Steel | |
1Each | |
Test Sieve | |
Stainless Steel | |
65mm |
For separation, fractioning, particle size determination | |
400 mm | |
36μm | |
ISO 3310/1 | |
Shaker AS 450 |