missing translation for 'onlineSavingsMsg'
Learn More

4-Vinylbenzeneboronic acid, 97%

Product Code. 11410138
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11410138

Brand: Thermo Scientific Alfa Aesar B23709.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2156-04-9 | C8H9BO2 | 147.97 g/mol

4-Vinylbenzeneboronic acid is widely used in Suzuki reaction. It is also used in the preparation of boronic acid containing polymers, which is used in potential biomedical applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Vinylbenzeneboronic acid is widely used in Suzuki reaction. It is also used in the preparation of boronic acid containing polymers, which is used in potential biomedical applications.

Solubility
Soluble in ethanol, dimethylsulfoxide, tetrahydrofuran and diethyl ether. Slightly soluble in water.

Notes
Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2156-04-9
Molecular Formula C8H9BO2
Molecular Weight (g/mol) 147.97
MDL Number MFCD00239441
InChI Key QWMJEUJXWVZSAG-UHFFFAOYSA-N
Synonym 4-vinylphenylboronic acid, 4-vinylbenzeneboronic acid, 4-vinylphenyl boronic acid, 4-ethenylphenyl boronic acid, 4-vinylboronic acid, p-styrylboronic acid, p-vinylphenylboronic acid, styrene-4-boronic acid, boronic acid, 4-ethenylphenyl, 4-styreneboronic acid
PubChem CID 2734393
IUPAC Name (4-ethenylphenyl)boronic acid
SMILES OB(O)C1=CC=C(C=C)C=C1

Specifications

Melting Point ∼190°C
Quantity 25 g
Beilstein 2829387
Solubility Information Soluble in ethanol,dimethylsulfoxide,tetrahydrofuran and diethyl ether. Slightly soluble in water.
Formula Weight 147.97
Percent Purity 97%
Chemical Name or Material 4-Vinylbenzeneboronic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.