Learn More
4-(Trifluoromethylthio)benzylamine, 95%, Thermo Scientific Chemicals
Product Code. 15424117
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15424117 1 g 1g
15434117 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15424117 Supplier Thermo Scientific Alfa Aesar Supplier No. H31515.03
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 128273-56-3
Molecular Formula C8H8F3NS
Molecular Weight (g/mol) 207.214
MDL Number MFCD00190129
InChI Key LACURGWEZCFLBO-UHFFFAOYSA-N
Synonym 4-trifluoromethylthio benzylamine, 4-trifluoromethyl thio phenyl methanamine, 4-trifluoromethylsulfanyl phenyl methanamine, 4-trifluoromethyl sulfanyl phenyl methanamine, 1-4-trifluoromethyl thio phenyl methanamine, 1-4-trifluoromethyl sulfanyl phenyl methanamine, acmc-1chvc, rarechem al bw 0450, 4-trifluomethylthio benzylamine
PubChem CID 2777875
IUPAC Name [4-(trifluoromethylsulfanyl)phenyl]methanamine
SMILES C1=CC(=CC=C1CN)SC(F)(F)F
Specifications
Density 1.31
Boiling Point 224°C to 225°C
Flash Point 80°C (176°F)
Refractive Index 1.504
Quantity 1 g
UN Number UN2735
Sensitivity Air Sensitive
Formula Weight 207.21
Percent Purity 95%
Chemical Name or Material 4-(Trifluoromethylthio)benzylamine