missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethoxy)benzoyl chloride, 98%

Product Code. p-3758884 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10591901

Brand: Thermo Scientific Acros 306350010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 36823-88-8 | C8H4ClF3O2 | 224.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36823-88-8
Molecular Formula C8H4ClF3O2
Molecular Weight (g/mol) 224.56
MDL Number MFCD00052329
InChI Key ZXKKOFJYPRJFIE-UHFFFAOYSA-N
Synonym 4-trifluoromethoxy benzoyl chloride, p-trifluoromethoxybenzoyl chloride, benzoyl chloride, 4-trifluoromethoxy, 4-trifluoromethoxy benzene-1-carbonyl chloride, 4-trifluoromethoxy benzoylchloride, 4-trifluoromethoxy-benzoyl chloride, p-trifluoromethoxy benzoyl chloride, acmc-209ip1, ksc495i1f, p-trifluoromethoxybenzoylchloride
PubChem CID 142110
IUPAC Name 4-(trifluoromethoxy)benzoyl chloride
SMILES C1=CC(=CC=C1C(=O)Cl)OC(F)(F)F

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Pink
Density 1.4200g/mL
Boiling Point 55°C to 56°C (2.0 mmHg)
Flash Point 32°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CF3OC6H4COCl
Refractive Index 1.4710 to 1.473
Quantity 1 g
Specific Gravity 1.42
Formula Weight 224.56
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 4-(Trifluoromethoxy)benzoyl chloride
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.