missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethoxy)aniline, TRC

Product Code. 30006378
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
50 g
100 g
250 g
missing translation for 'unitSize'
100g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30006378 50 g 50g
30132861 100 g 100g
30135856 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 30006378 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCT79070050G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 461-82-5
Formula Weight 177.0401
InChI Formula InChI=1S/C7H6F3NO/c8-7(9,10)12-6-3-1-5(11)2-4-6/h1-4H,11H2
IUPAC Name 4-(trifluoromethoxy)aniline
Molecular Formula C7 H6 F3 N O
Molecular Weight (g/mol) 177.12
SMILES Nc1ccc(OC(F)(F)F)cc1
Synonym 4-(Trifluoromethoxy)-benzenamine, p-Anisidine, α,α,α-trifluoro- (6CI,7CI,8CI), (4-(Trifluoromethoxy)phenyl)amine, 1-Amino-4-(trifluoromethoxy)benzene, 4-(Trifluoromethoxy)aniline, 4-(Trifluoromethoxy)benzenamine, 4-Trifluoromethoxybenzeneamine, 4-[(Trifluoromethyl)oxy]aniline, p-(Trifluoromethoxy)aniline, p-Trifluoromethoxyphenylamine, α,α,α-Trifluoro-p-anisidine
Chemical Name or Material 4-(Trifluoromethoxy)aniline

Specifications

Quantity 50 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.