Learn More
4-Tolyl-d4-sulfonamide, TRC
Product Code. 30449797
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30449797 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30449797 Supplier Toronto Research Chemicals Supplier No. TRCT53663225MG
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
Add to basket
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 4-Tolyl-d4-sulfonamide
CAS 1219795-34-2
Molecular Formula C7 2H4 H5 N O2 S
Quantity 25 mg
Synonym p-Toluene-2,3,5,6-d4-sulfonamide, 4-Tolyl-d4-sulfonamide, 4-Methylbenzene-2,3,5,6-d4-sulfonamide, 4-Methylbenzenesulfonamide-2,3,5,6-D4, p-Tolylsulfonamide-d4
InChI Formula InChI=1S/C7H9NO2S/c1-6-2-4-7(5-3-6)11(8,9)10/h2-5H,1H3,(H2,8,9,10)/i2D,3D,4D,5D
SMILES [2H]c1c([2H])c(c([2H])c([2H])c1C)S(=O)(=O)N
IUPAC Name 2,3,5,6-tetradeuterio-4-methylbenzenesulfonamide
Molecular Weight (g/mol) 175.24
Formula Weight 175.0605