Learn More
4-tert-Butylbenzoic-d13 Acid, CDN
Specifications
Specifications
| Chemical Name or Material | 4-tert-Butylbenzoic Acid-D13 |
| CAS | 2733271-85-5 |
| Molecular Formula | C11 D13 H O2 |
| Quantity | 0.1 g |
| Synonym | 4-tert-Butylbenzoic-d13 Acid, 4-(tert-Butyl-d9)benzoic-2,3,5,6-d4 acid, 4-[1,1-Di(methy-d3)lethyl-1,2,2,2-d4]benzoic-2,3,5,6-d4 acid, 4-tert-Butylbenzoic acid-D13 |
| InChI Formula | InChI=1 S/C11H14O2/c1-11(2,3)9-6-4-8(5-7-9)10(12)13/h4-7 H,1-3H3,(H,12,13)/i1D3,2D3,3D3,4 D,5 D,6 D,7 D |
| SMILES | [2 H]c1c([2 H])c(c([2 H])c([2 H])c1C(=O)O)C(C([2 H])([2 H])[2 H])(C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | 2,3,5,6-tetradeuterio-4-[1,1,1,3,3,3-hexadeuterio-2-(trideuteriomethyl)propan-2-yl]benzoic acid |
| Molecular Weight (g/mol) | 191.3078 |
| Formula Weight | 191.181 g/mol |
| Show More |