Learn More
4-tert-Butylbenzenesulfonamide, TRC
Product Code. 30348673
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30348673 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30348673 Supplier Toronto Research Chemicals Supplier No. TRCB69073510G
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 4-(tert-butyl)benzenesulfonamide
CAS 6292-59-7
Molecular Formula C10 H15 N O2 S
Quantity 10 g
Synonym Benzenesulfonamide, 4-(1,1-dimethylethyl)-, Benzenesulfonamide, p-tert-butyl- (6CI,8CI), 4-(1,1-Dimethylethyl)benzenesulfonamide, 4-tert-Butylbenzenesulfonamide, NSC 9911, p-(tert-Butyl)phenylsulfonamide, p-tert-Butylbenzenesulfonamide, (4-(tert-butyl)benzenesulfonamide), Bosentan Related Compound E
InChI Formula InChI=1S/C10H15NO2S/c1-10(2,3)8-4-6-9(7-5-8)14(11,12)13/h4-7H,1-3H3,(H2,11,12,13)
SMILES CC(C)(C)c1ccc(cc1)S(=O)(=O)N
IUPAC Name 4-tert-butylbenzenesulfonamide
Molecular Weight (g/mol) 213.3
Formula Weight 213.0823