Learn More
4-tert-Butyl-d9-phenol-2,3,5,6-d4, TRC
Specifications
Specifications
| Chemical Name or Material | 4-tert-Butyl-d9-phenol-2,3,5,6-d4 |
| CAS | 225386-58-3 |
| Molecular Formula | C10 D13 H O |
| Quantity | 10 mg |
| Synonym | Phen-2,3,5,6-d4-ol, 4-[1,1-di(methyl-d3)ethyl-2,2,2-d3]- (9CI) |
| InChI Formula | InChI=1S/C10H14O/c1-10(2,3)8-4-6-9(11)7-5-8/h4-7,11H,1-3H3/i1D3,2D3,3D3,4D,5D,6D,7D |
| SMILES | [2H]c1c([2H])c(c([2H])c([2H])c1O)C(C([2H])([2H])[2H])(C([2H])([2H])[2H])C([2H])([2H])[2H] |
| IUPAC Name | 2,3,5,6-tetradeuterio-4-[1,1,1,3,3,3-hexadeuterio-2-(trideuteriomethyl)propan-2-yl]phenol |
| Molecular Weight (g/mol) | 163.3 |
| Formula Weight | 163.1861 |