missing translation for 'onlineSavingsMsg'
Learn More

4-(tert-Butyl)-2-chloroaniline, 95%, Thermo Scientific™

Product Code. p-3110571
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10310653 1 g Amber Glass Bottle 1g
10609724 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10310653 Supplier Thermo Scientific Maybridge Supplier No. RF01508DA
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 42265-67-8
Molecular Formula C10H14ClN
Molecular Weight (g/mol) 183.68
MDL Number MFCD00052272
InChI Key MFEZEFHCSBXPPO-UHFFFAOYSA-N
Synonym 4-tert-butyl-2-chloroaniline, 4-t-butyl-2-chloroaniline, 2-chloro-4-tert-butylaniline, 4-tert-butyl-2-chloro-aniline, aniline, 4-tert-butyl-2-chloro, 4-tert-butyl-2-chlorophenylamine, benzenamine,2-chloro-4-1,1-dimethylethyl
PubChem CID 521094
IUPAC Name 4-tert-butyl-2-chloroaniline
SMILES CC(C)(C)C1=CC(Cl)=C(N)C=C1

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 183.68
Percent Purity 95%
Chemical Name or Material 4-(tert-Butyl)-2-chloroaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.