Learn More
4-[(tert-Butoxycarbonyl)oxy]-2-methylphenol, TRC
Specifications
Specifications
| Chemical Name or Material | 4-[(tert-Butoxycarbonyl)oxy]-2-methylphenol |
| CAS | 457634-20-7 |
| Molecular Formula | C12 H16 O4 |
| Quantity | 2 g |
| InChI Formula | InChI=1S/C12H16O4/c1-8-7-9(5-6-10(8)13)15-11(14)16-12(2,3)4/h5-7,13H,1-4H3 |
| SMILES | Cc1cc(OC(=O)OC(C)(C)C)ccc1O |
| IUPAC Name | tert-butyl (4-hydroxy-3-methylphenyl) carbonate |
| Molecular Weight (g/mol) | 224.25 |
| Formula Weight | 224.1049 |