missing translation for 'onlineSavingsMsg'
Learn More

[4-(Pyrid-2-yloxy)phenyl]methanol, 97%, Thermo Scientific™

Product Code. p-3484537
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10539223 250 mg Amber Glass Bottle 250mg
10367644 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10539223 Supplier Thermo Scientific Maybridge Supplier No. CC48909CB
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 194017-70-4
Molecular Formula C12H11NO2
Molecular Weight (g/mol) 201.23
MDL Number MFCD08690278
InChI Key ZDTXOQRCIVCMLT-UHFFFAOYSA-N
Synonym 4-pyrid-2-yloxy phenyl methanol, 4-pyridin-2-yloxy phenyl methanol, 4-2-pyridyloxy phenyl methanol, 4-pyridin-2-yl oxy phenyl, 2-4-hydroxymethylphenoxy pyridine, 4-2-pyridyloxy phenyl methan-1-ol, 4-pyridin-2-yl oxy phenyl methanol
PubChem CID 17859715
SMILES OCC1=CC=C(OC2=CC=CC=N2)C=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
IUPAC Name (4-pyridin-2-yloxyphenyl)methanol
Formula Weight 201.22
Percent Purity 97%
Chemical Name or Material [4-(Pyrid-2-yloxy)phenyl]methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.