missing translation for 'onlineSavingsMsg'
Learn More

4-Phenyl-2-(2-pyridyl)thiazole-5-carboxylic acid, 97%, Thermo Scientific™

Product Code. 15409365
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15409365 250 mg 250mg
15499355 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15409365 Supplier Thermo Scientific Alfa Aesar Supplier No. H51850.MD
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1301738-68-0
Molecular Formula C15H10N2O2S
Molecular Weight (g/mol) 282.32
MDL Number MFCD16251584
InChI Key AFWYDGNOZTZZDG-UHFFFAOYSA-N
Synonym 4-phenyl-2-2-pyridyl thiazole-5-carboxylic acid, 4-phenyl-2-pyridin-2-yl thiazole-5-carboxylic acid, 4-phenyl-2-pyridin-2-yl-1,3-thiazole-5-carboxylic acid
PubChem CID 73012805
IUPAC Name 4-phenyl-2-pyridin-2-yl-1,3-thiazole-5-carboxylic acid
SMILES OC(=O)C1=C(N=C(S1)C1=CC=CC=N1)C1=CC=CC=C1

Specifications

Quantity 250 mg
Formula Weight 282.32
Percent Purity 97%
Chemical Name or Material 4-Phenyl-2-(2-pyridyl)thiazole-5-carboxylic acid
compliance-icons
Product Identifier
  • 4-Phenyl-2-(2-pyridyl)thiazole-5-carboxylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 4-Phenyl-2-(2-pyridyl)thiazole-5-carboxylic
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.