missing translation for 'onlineSavingsMsg'
Learn More

4-(Pentafluorothio)benzoyl chloride, 97%, Thermo Scientific™

Catalog Number p-7032916
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
197384-98-8
C7H4ClF5OS
266.61
MFCD03788523
NNFDYUZNKYWPGP-UHFFFAOYSA-N
4-pentafluorosulfanyl benzoyl chloride, 4-pentafluorothio benzoyl chloride, sulfur, 4-chlorocarbonyl phenyl pentafluoro-, oc-6-21-9ci, 4-pentafluoro-??-sulfanyl benzoyl chloride, 4-pentafluorosulfanylbenzoic chloride, 4-pentafluorosulphanylbenzoyl chloride, 4-pentafluorosulphanyl benzoyl chloride, 4-chlorocarbonyl phenylsulphur pentafluoride, 4-pentafluoro-$l^ 6-sulfanyl benzoyl chloride, 4-pentafluoro-lambda6-sulfanyl benzoyl chloride
2779215
4-(pentafluoro-$l^{6}-sulfanyl)benzoyl chloride
FS(F)(F)(F)(F)C1=CC=C(C=C1)C(Cl)=O
This item is not returnable. View return policy
197384-98-8
C7H4ClF5OS
266.61
MFCD03788523
NNFDYUZNKYWPGP-UHFFFAOYSA-N
4-pentafluorosulfanyl benzoyl chloride, 4-pentafluorothio benzoyl chloride, sulfur, 4-chlorocarbonyl phenyl pentafluoro-, oc-6-21-9ci, 4-pentafluoro-??-sulfanyl benzoyl chloride, 4-pentafluorosulfanylbenzoic chloride, 4-pentafluorosulphanylbenzoyl chloride, 4-pentafluorosulphanyl benzoyl chloride, 4-chlorocarbonyl phenylsulphur pentafluoride, 4-pentafluoro-$l^ 6-sulfanyl benzoyl chloride, 4-pentafluoro-lambda6-sulfanyl benzoyl chloride
2779215
4-(pentafluoro-$l^{6}-sulfanyl)benzoyl chloride
FS(F)(F)(F)(F)C1=CC=C(C=C1)C(Cl)=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
UN Number UN3265
Sensitivity Moisture sensitive
Formula Weight 266.61
Percent Purity 97%
Chemical Name or Material 4-(Pentafluorothio)benzoyl chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'