Learn More
4-Oxopentanoic-13C3 Acid, TRC
Specifications
Specifications
| Chemical Name or Material | 4-Oxopentanoic-13C3 Acid |
| CAS | 1391051-93-6 |
| Molecular Formula | C213C3H8O3 |
| Quantity | 25 mg |
| Synonym | 3-Acetylpropionic-13C3 Acid, 3-Oxobutanecarboxylic-13C3 Acid, 4-Ketovaleric-13C3 Acid, 4-Oxopentanoic-13C3 Acid, 4-Oxovaleric-13C3 Acid, Laevulinic-13C3 Acid, Levulic-13C3 Acid, NSC 3716-13C3, β-Acetylpropionic-13C3 Acid, γ-Ketovaleric-13C3 Acid, Levulinic-13C3 Acid |
| InChI Formula | InChI=1S/C5H8O3/c1-4(6)2-3-5(7)8/h2-3H2,1H3,(H,7,8)/i1+1,2+1,4+1 |
| SMILES | [13CH3][13C]([13CH2]CC(O)=O)=O |
| Molecular Weight (g/mol) | 119.09 |
| Formula Weight | 119.0574 |