missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies 4-Octyl Itaconate
Description
- 4-Octyl Itaconate is a cell-permeable derivative of itaconate, a metabolite produced during the Krebs cycle
- It is a potent activator of nuclear factor erythroid 2-related factor 2 (Nrf2) via alkylation of the inhibitor of Nrf2 (Keap1), thereby promoting Nrf2-mediated transcriptional activation and downstream anti-oxidant and anti-inflammatory properties (Mills et al.; Tang et al.)
- The itaconate pathway (IRG1-itaconate-SDH) plays a pivotal role in up-regulating tolerance and inhibiting the induction of trained immunity (Domínguez-Andrés et al.)
- Counteracts the proinflammatory signals of succinate by inhibiting succinate dehydrogenase (SDH) and activating Nrf2 to limit inflammation in mouse and human macrophages (Lampropoulou et al.; Mills et al.)
- Activates Nrf2 signaling in human macrophages, which has been shown to inhibit the production of pro-inflammatory cytokines including TNF-α, IL-1β, and IL-6 (Tang et al.)
Specifications
Specifications
| CAS | 3133-16-2 |
| Synonym | 2-methylene-butanedioic acid; 4-octyl ester |
| Quantity | 250 mg |
| Molecular Formula | C13H22O4 |
| Purity | ≥98% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?