Learn More
4-O-Acetyl-3,6-di-O-(3,4,6-tri-O-acetyl-Beta-D-mannopyranosyl-ethylidyne)-1,2-O-ethylidene-Beta-D-mannopyranose, TRC
Specifications
Specifications
| Chemical Name or Material | 4-O-Acetyl-3,6-di-O-(3,4,6-tri-O-acetyl-beta-D-mannopyranosyl-ethylidyne)-1,2-O-ethylidene-beta-D-mannopyranose |
| CAS | 230963-27-6 |
| Molecular Formula | C38H52O25 |
| Quantity | 5 mg |
| Synonym | O-3,4,6-Tri-O-acetyl-beta-D-mannopyranosylethylidyne-(1-23)-O-[3,4,6-tri-O-acetyl-beta-D-mannopyranosylethylidyne-(1-26)]-1,2-O-ethylidene-beta-D-mannopyranose Acetate |
| InChI Formula | InChI=1S/C38H52O25/c1-14(39)46-11-22-25(49-16(3)41)28(52-19(6)44)32-35(57-22)62-37(9,60-32)48-13-24-27(51-18(5)43)30(31-34(56-24)55-21(8)54-31)59-38(10)61-33-29(53-20(7)45)26(50-17(4)42)23(12-47-15(2)40)58-36(33)63-38/h21-36H,11-13H2,1-10H3 |
| SMILES | CC1O[C@@H]2O[C@H](COC3(C)O[C@@H]4O[C@H](COC(=O)C)[C@@H](OC(=O)C)[C@H](OC(=O)C)[C@@H]4O3)[C@@H](OC(=O)C)[C@H](OC5(C)O[C@@H]6O[C@H](COC(=O)C)[C@@H](OC(=O)C)[C@H](OC(=O)C)[C@@H]6O5)[C@@H]2O1 |
| IUPAC Name | [6,7-diacetyloxy-2-[[6-acetyloxy-7-[[6,7-diacetyloxy-5-(acetyloxymethyl)-2-methyl-5,6,7,7a-tetrahydro-3aH-[1,3]dioxolo[4,5-b]pyran-2-yl]oxy]-2-methyl-5,6,7,7a-tetrahydro-3aH-[1,3]dioxolo[4,5-b]pyran-5-yl]methoxy]-2-methyl-5,6,7,7a-tetrahydro-3aH-[1,3]dioxolo[4,5-b]pyran-5-yl]methyl acetate |
| Molecular Weight (g/mol) | 908.8 |