Learn More
4-Nonylphenoxyacetic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | 4-Nonylphenoxyacetic Acid |
| CAS | 3115-49-9 |
| Molecular Formula | C17 H26 O3 |
| Quantity | 100 mg |
| Synonym | 4-Nonylphenoxyacetic Acid, Acetic acid','2-(4-nonylphenoxy)- (ACI), 2-(4-Nonylphenoxy)acetic acid (ACI), Acetic acid','(4-nonylphenoxy)- (9CI), Acetic acid','(p-nonylphenoxy)- (6CI','7CI','8CI), (p-Nonylphenoxy)acetic acid, 4-Nonylphenoxyacetic acid |
| InChI Formula | InChI=1S/C17H26O3/c1-2-3-4-5-6-7-8-9-15-10-12-16(13-11-15)20-14-17(18)19/h10-13H,2-9,14H2,1H3,(H,18,19) |
| SMILES | CCCCCCCCCc1ccc(OCC(=O)O)cc1 |
| IUPAC Name | 2-(4-nonylphenoxy)acetic acid |
| Molecular Weight (g/mol) | 278.39 |
| Formula Weight | 278.1882 |